Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152695.5 | NP_689908.3 | 3 | 689 | ||
XM_011531312.2 | XP_011529614.1 | 4 | 759 | |||
XM_011531313.2 | XP_011529615.1 | 2 | 1305 | |||
XM_011531314.2 | XP_011529616.1 | 4 | 1446 | |||
XM_017029350.1 | XP_016884839.1 | 3 | 637 | |||
XM_017029351.1 | XP_016884840.1 | 3 | 734 | |||
GenBank mRNA | AK314344.1 | - | 3 | 666 | ||
AL833109.1 | - | 3 | 647 | |||
AL833407.1 | - | 3 | 665 | |||
AY280801.1 | - | 3 | 665 | |||
BC060768.1 | - | 3 | 664 | 30346713 | ||
BC065938.1 | - | 4 | 736 |