Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105565.2 | NP_001099035.2 | 1 | 639 | ||
XM_005273819.3 | XP_005273876.1 | 2 | 1441 |