Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244889.1 | NP_001231818.1 | 15 | 3018 | ||
NM_170744.4 | NP_734465.2 | 16 | 3051 | |||
XM_011539453.1 | XP_011537755.1 | 17 | 2600 | |||
XM_017015834.1 | XP_016871323.1 | 17 | 2610 | |||
XM_017015835.1 | XP_016871324.1 | 16 | 3403 | |||
GenBank mRNA | AB096256.1 | - | 16 | 3006 | ||
AK022859.1 | - | 7 | 1066 | |||
AK094595.1 | - | 9 | 1521 | |||
AK128132.1 | - | 16 | 3051 | |||
AY126437.1 | - | 15 | 2626 | |||
AY358351.1 | - | 16 | 2997 |