Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282941.1 | NP_001269870.1 | 2 | 438 | ||
NM_145307.3 | NP_660350.2 | 2 | 438 | |||
XM_011539456.2 | XP_011537758.1 | 2 | 454 | |||
XM_011539457.2 | XP_011537759.1 | 2 | 454 | |||
XM_011539460.2 | XP_011537762.1 | 2 | 454 | |||
XM_017015842.1 | XP_016871331.1 | 2 | 415 | |||
XM_017015843.1 | XP_016871332.1 | 2 | 454 | |||
XM_017015844.1 | XP_016871333.1 | 2 | 454 | |||
GenBank mRNA | AY150309.1 | - | 2 | 282 | ||
BC025765.1 | - | 2 | 282 | |||
BC141821.1 | - | 2 | 438 | |||
BC142725.1 | - | 2 | 438 | |||
BX113509.1 | - | 2 | 390 |