Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152721.5 | NP_689934.2 | 2 | 298 | ||
XM_011525875.2 | XP_011524177.1 | 2 | 319 | |||
GenBank mRNA | AY599248.1 | - | 2 | 296 | ||
BC096744.1 | - | 2 | 291 | 5260167 | ||
DB480182.1 | - | 2 | 298 |