Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF438 SIRNA Assays ›
Gene Symbol:
ZNF438
Gene Name:
zinc finger protein 438
Gene Aliases:
bA330O11.1
Chromosome Location:
Chr.10: 30844634 - 31032406 on Build GRCh38
UniGene ID:
Hs.660642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47911
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF438
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143766.1
NP_001137238.1
7
1795
NM_001143767.1
NP_001137239.1
5
1608
NM_001143768.1
NP_001137240.1
6
1666
NM_001143769.1
NP_001137241.1
7
1920
NM_001143770.1
NP_001137242.1
8
1846
NM_001143771.1
NP_001137243.1
7
1817
NM_182755.2
NP_877432.1
7
1747
NR_026560.1
6
830
XM_006717393.3
XP_006717456.1
5
1562
XM_006717396.3
XP_006717459.1
4
1496
XM_006717398.3
XP_006717461.1
8
1787
XM_006717399.3
XP_006717462.1
7
1720
XM_011519371.2
XP_011517673.1
6
3770
XM_011519372.2
XP_011517674.1
6
1797
XM_011519373.2
XP_011517675.1
5
3712
XM_011519374.2
XP_011517676.1
3
1539
XM_011519376.2
XP_011517678.1
8
1872
XM_011519377.2
XP_011517679.1
8
1868
XM_017015863.1
XP_016871352.1
3
1362
XM_017015864.1
XP_016871353.1
8
4006
XM_017015865.1
XP_016871354.1
7
3947
XM_017015866.1
XP_016871355.1
7
1837
XM_017015867.1
XP_016871356.1
6
3752
XM_017015868.1
XP_016871357.1
5
2058
XM_017015869.1
XP_016871358.1
5
3693
XM_017015870.1
XP_016871359.1
6
2592
XM_017015871.1
XP_016871360.1
5
2052
XM_017015872.1
XP_016871361.1
6
3750
XM_017015873.1
XP_016871362.1
7
3927
XM_017015874.1
XP_016871363.1
5
1613
XM_017015875.1
XP_016871364.1
7
816
XM_017015876.1
XP_016871365.1
9
1565
XM_017015877.1
XP_016871366.1
8
995
XM_017015878.1
XP_016871367.1
7
922
XM_017015879.1
XP_016871368.1
7
1002
XM_017015880.1
XP_016871369.1
6
805
XM_017015881.1
XP_016871370.1
5
743
XM_017015882.1
XP_016871371.1
6
850
GenBank mRNA
AF428258.1
-
6
1642
AF440405.1
-
1
107
AK057323.1
-
6
818
AK131357.1
-
7
1920
AK292730.1
-
7
1713
AK294253.1
-
8
741
AL833056.1
-
2
1356
BC101622.1
-
3
1372
8069128
BC104757.1
-
3
1372
8143760
BC143462.1
-
4
1471
BC143464.1
-
4
1551
DQ356011.1
-
7
1741
DQ356012.1
-
7
1732
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0