Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47935
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001026383.2
NP_001021554.1
2
624
NM_001321115.1
NP_001308044.1
2
858
NM_145023.5
NP_659460.3
2
858
NR_109826.1
2
624
NR_109827.1
2
858
XM_006717505.1
XP_006717568.1
2
566
XM_006717506.1
XP_006717569.1
2
566
XM_011519663.1
XP_011517965.1
2
566
XM_011519664.1
XP_011517966.1
2
566
XM_011519666.1
XP_011517968.1
2
566
XM_011519667.1
XP_011517969.1
2
566
XM_011519668.1
XP_011517970.1
2
566
XM_011519669.1
XP_011517971.1
2
566
XM_011519670.1
XP_011517972.1
2
566
XM_011519671.1
XP_011517973.1
2
566
XM_011519672.1
XP_011517974.1
2
566
XM_011519673.1
XP_011517975.1
2
566
XM_011519674.1
XP_011517976.1
2
566
XM_011519675.1
XP_011517977.1
2
566
XM_011519676.1
XP_011517978.1
2
566
XM_011519677.1
XP_011517979.1
2
566
XM_011519678.1
XP_011517980.1
2
566
XM_011519679.1
XP_011517981.1
2
566
XM_011519684.2
XP_011517986.1
2
566
XM_011519686.2
XP_011517988.1
2
566
XM_011519688.2
XP_011517990.1
2
566
XM_017016638.1
XP_016872127.1
2
566
XM_017016639.1
XP_016872128.1
2
566
XM_017016640.1
XP_016872129.1
2
566
XM_017016644.1
XP_016872133.1
2
566
XM_017016647.1
XP_016872136.1
2
566
XM_017016650.1
XP_016872139.1
2
566
XM_017016651.1
XP_016872140.1
2
566
GenBank mRNA
AK057324.1
-
2
566
BC022020.2
-
2
764
4825109
BC042672.1
-
2
549
5166976
BX647771.1
-
2
554
EF656618.1
-
1
279
EF656619.1
-
1
279
EF656620.1
-
1
279
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0