Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 23 | 4668 | ||
GenBank mRNA | AB209312.1 | - | 23 | 4460 | ||
LM994822.1 | - | 23 | 4546 |
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 15 | 3610 | ||
GenBank mRNA | AF462447.1 | - | 15 | 3567 | ||
AK294708.1 | - | 15 | 3539 | |||
BC031522.2 | - | 15 | 3596 | 3869893 | ||
BX647399.1 | - | 15 | 3439 | |||
D16305.1 | - | 15 | 3191 | |||
L20046.1 | - | 15 | 3232 | |||
LM994821.1 | - | 11 | 2680 | |||
X69978.1 | - | 15 | 3381 |