Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286574.1 | NP_001273503.1 | 6 | 783 | ||
NM_145028.4 | NP_659465.2 | 6 | 864 | |||
XM_011514381.2 | XP_011512683.1 | 6 | 732 | |||
XM_011514382.2 | XP_011512684.1 | 6 | 863 | |||
XM_017010435.1 | XP_016865924.1 | 7 | 2338 | |||
GenBank mRNA | BC033033.2 | - | 6 | 814 | 5268250 | |
CR735810.1 | - | 6 | 677 |