Gene Symbol: PXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183373.3 | NP_899229.2 | 4, 5 | 1057 | ||
XM_011514393.2 | XP_011512695.1 | 6, 7 | 1036 | |||
GenBank mRNA | AK024828.1 | - | 4, 5 | 530 | ||
AK289598.1 | - | 4, 5 | 758 | |||
AL832792.1 | - | 4, 5 | 603 | |||
BC056908.1 | - | 4, 5 | 747 | 6158428 |