Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWISTNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002926.1 | NP_001002926.1 | 2, 3 | 416 | ||
GenBank mRNA | AK301329.1 | - | 2, 3 | 440 | ||
BC014574.1 | - | 2, 3 | 418 | 3685950 | ||
BC056267.1 | - | 2, 3 | 430 | |||
BC130298.1 | - | 2, 3 | 415 | 40146329 | ||
BC130300.1 | - | 2, 3 | 414 | 40146331 |