Search Thermo Fisher Scientific
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175061.3 | NP_778231.2 | 1, 2 | 354 | ||
XM_006715656.1 | XP_006715719.1 | 1, 2 | 168 | |||
GenBank mRNA | AK091311.1 | - | 1, 2 | 279 | ||
AY372319.1 | - | 1, 2 | 132 | |||
BC042441.1 | - | 1, 2 | 142 | 4831063 | ||
BC047229.1 | - | 1, 2 | 141 | |||
DA864624.1 | - | 1, 2 | 354 |