Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 2, 3 | 478 | ||
XM_005249652.3 | XP_005249709.1 | 2, 3 | 421 | |||
GenBank mRNA | AK294982.1 | - | 2, 3 | 208 | ||
AL834391.1 | - | 2, 3 | 412 | |||
AY054121.1 | - | 2, 3 | 274 | |||
BC062423.1 | - | 2, 3 | 229 | |||
BI560774.1 | - | 2, 3 | 482 | |||
DT217816.1 | - | 2, 3 | 413 |