Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317721.1 | NP_001304650.1 | 8 | 1210 | ||
NM_152892.2 | NP_690852.1 | 8 | 1192 | |||
XM_017011845.1 | XP_016867334.1 | 8 | 1147 | |||
GenBank mRNA | AB209379.1 | - | 5 | 1559 | ||
AK290967.1 | - | 8 | 1091 | |||
AK313771.1 | - | 8 | 1075 | |||
AL133057.1 | - | 8 | 1142 | |||
BC009436.1 | - | 7 | 883 | |||
BC018769.1 | - | 7 | 722 | |||
BC030547.1 | - | 8 | 1132 | 5170588 |