Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308360.1 | NP_001295289.1 | 10 | 1479 | ||
NM_170776.4 | NP_740746.4 | 11 | 1618 | |||
XM_005255842.3 | XP_005255899.1 | 12 | 1812 | |||
XM_011522951.2 | XP_011521253.1 | 11 | 1566 | |||
XM_011522953.2 | XP_011521255.1 | 9 | 1296 | |||
GenBank mRNA | AB049169.1 | - | 11 | 1561 | ||
AK172801.1 | - | 11 | 1619 | |||
AK301258.1 | - | 10 | 1416 | |||
AK310311.1 | - | 5 | 625 | |||
AY140959.1 | - | 11 | 1515 | |||
BC064508.1 | - | 11 | 1604 | 6424664 |
Set Membership: |
![]() |