Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173561.2 | NP_775832.2 | 5, 6 | 1147 | ||
GenBank mRNA | AY510109.1 | - | 4, 5 | 1048 | ||
BC035284.1 | - | 5, 6 | 1137 | 5189247 | ||
BM312066.1 | - | 2, 3 | 189 | |||
JF681947.1 | - | 5, 6 | 1165 |