Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ESR2 SIRNA Assays ›
Gene Symbol:
ESR2
Gene Name:
estrogen receptor 2
Gene Aliases:
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
Chromosome Location:
Chr.14: 64226712 - 64338631 on Build GRCh38
UniGene ID:
Hs.660607
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4827
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ESR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040275.1
NP_001035365.1
2, 3
823
NM_001214902.1
NP_001201831.1
1, 2
445
NM_001271876.1
NP_001258805.1
1, 2
445
NM_001271877.1
NP_001258806.1
1, 2
445
NM_001291712.1
NP_001278641.1
7, 8
2083
NM_001291723.1
NP_001278652.1
2, 3
1098
NM_001437.2
NP_001428.1
2, 3
823
NR_073496.1
2, 3
1098
XM_017021079.1
XP_016876568.1
3, 4
1487
XM_017021080.1
XP_016876569.1
3, 4
2296
XM_017021081.1
XP_016876570.1
3, 4
1392
XM_017021082.1
XP_016876571.1
3, 4
1244
XM_017021083.1
XP_016876572.1
2, 3
661
XM_017021084.1
XP_016876573.1
3, 4
1487
GenBank mRNA
AB006589.1
-
7, 8
1630
AB006590.1
-
1, 2
453
AB209620.1
-
1, 2
451
AF051427.1
-
2, 3
773
AF051428.1
-
2, 3
773
AF060555.1
-
2, 3
825
AF074599.1
-
1, 2
183
AF124790.1
-
1, 2
406
AK292370.1
-
2, 3
660
BC024181.2
-
2, 3
640
4825937
DQ777076.1
-
1, 2
355
DQ777077.1
-
1, 2
355
DQ838582.1
-
1, 2
355
DQ838583.1
-
1, 2
355
HQ692820.1
-
1, 2
355
HQ692821.1
-
1, 2
355
KJ534833.1
-
1, 2
355
X99101.1
-
1, 2
214
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012754, XM_006240221, AB012721, AF042060, AB190770, and others.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
ovarian follicle development
behavioral fear response
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
signal transduction
cell-cell signaling
learning or memory
response to water deprivation
response to activity
response to insecticide
cerebellum development
amygdala development
hypothalamus development
negative regulation of cell growth
intracellular estrogen receptor signaling pathway
prostate gland development
response to nutrient levels
response to testosterone
response to genistein
cellular response to drug
positive regulation of apoptotic process
regulation of neuron apoptotic process
response to estrogen
response to human chorionic gonadotropin
estrous cycle
response to ethanol
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of smooth muscle cell proliferation
negative regulation of epithelial cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
Sertoli cell development
Sertoli cell proliferation
positive regulation of ERK1 and ERK2 cascade
cellular response to magnetism
cellular response to estradiol stimulus
response to dexamethasone
extracellular negative regulation of signal transduction
negative regulation of neuron death
response to salt
response to bisphenol A
negative regulation of feeding behavior
negative regulation of reactive oxygen species metabolic process
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
protein binding
drug binding
zinc ion binding
enzyme binding
estrogen receptor activity
estrogen response element binding
RNA polymerase II transcription factor activity, estrogen-activated sequence-specific DNA binding
peroxisome proliferator activated receptor binding
receptor antagonist activity
estradiol binding
steroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0