Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037729.1 | NP_001032818.1 | 2 | 146 | ||
GenBank mRNA | DQ012017.1 | - | 2 | 146 |