Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010874.4 | NP_001010874.2 | 1 | 288 | ||
XM_005265662.4 | XP_005265719.1 | 1 | 337 | |||
XM_005265663.2 | XP_005265720.1 | 1 | 288 | |||
XM_005265664.2 | XP_005265721.1 | 1 | 288 | |||
XM_005265665.3 | XP_005265722.1 | 1 | 288 | |||
XM_017007959.1 | XP_016863448.1 | 1 | 288 | |||
XM_017007960.1 | XP_016863449.1 | 1 | 288 | |||
GenBank mRNA | AK290606.1 | - | 1 | 284 | ||
AL833108.1 | - | 1 | 289 | |||
BC072463.1 | - | 1 | 288 | 4245690 | ||
BX647590.1 | - | 1 | 225 |