Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152760.2 | NP_689973.2 | 2 | 255 | ||
XM_006718488.3 | XP_006718551.1 | 2 | 558 | |||
XM_017017470.1 | XP_016872959.1 | 2 | 533 | |||
XM_017017471.1 | XP_016872960.1 | 3 | 717 | |||
XM_017017472.1 | XP_016872961.1 | 3 | 463 | |||
XM_017017473.1 | XP_016872962.1 | 3 | 463 | |||
GenBank mRNA | AK055496.1 | - | 2 | 215 | ||
AK289587.1 | - | 2 | 221 | |||
AK293827.1 | - | 2 | 242 | |||
AK299512.1 | - | 2 | 231 | |||
AK309435.1 | - | 2 | 221 | |||
AK311588.1 | - | 3 | 381 | |||
AL833261.1 | - | 2 | 241 | |||
BC040981.2 | - | 2 | 231 | 4797976 | ||
BC045563.1 | - | 2 | 204 | 4815203 |