Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ETS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256295.1 | NP_001243224.1 | 10 | 1624 | ||
NM_005239.5 | NP_005230.1 | 9 | 1454 | |||
XM_005260935.1 | XP_005260992.1 | 9 | 1527 | |||
XM_017028290.1 | XP_016883779.1 | 9 | 4124 | |||
GenBank mRNA | AK096841.1 | - | 10 | 1624 | ||
AK315563.1 | - | 9 | 1357 | |||
BC017040.1 | - | 9 | 1326 | 3852274 | ||
BC042954.2 | - | 9 | 1356 | 5276704 | ||
BT006838.1 | - | 8 | 1164 | |||
J04102.1 | - | 9 | 1455 | |||
M11922.1 | - | 2 | 195 |