Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207340.1 | NP_997223.1 | 3 | 1132 | ||
GenBank mRNA | AK301787.1 | - | 3 | 1166 | ||
AY358448.1 | - | 3 | 1132 | |||
BC005015.1 | - | 2 | 546 | |||
BC057833.1 | - | 3 | 961 | 30343331 |