Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152769.2 | NP_689982.3 | 6 | 805 | ||
XM_017026555.1 | XP_016882044.1 | 6 | 924 | |||
XM_017026556.1 | XP_016882045.1 | 6 | 779 | |||
XM_017026557.1 | XP_016882046.1 | 5 | 1267 | |||
XM_017026558.1 | XP_016882047.1 | 5 | 410 | |||
XM_017026559.1 | XP_016882048.1 | 6 | 924 | |||
XM_017026560.1 | XP_016882049.1 | 6 | 924 | |||
GenBank mRNA | BC028156.1 | - | 6 | 656 | 5241080 | |
BM743507.1 | - | 2 | 178 |