Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCO4 SIRNA Assays ›
Gene Symbol:
TMCO4
Gene Name:
transmembrane and coiled-coil domains 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 19682213 - 19800385 on Build GRCh38
UniGene ID:
Hs.656313
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48612
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCO4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181719.4
NP_859070.3
14
1608
XM_005245822.4
XP_005245879.1
14
1624
XM_005245824.4
XP_005245881.1
14
1688
XM_005245825.4
XP_005245882.1
13
1489
XM_005245827.3
XP_005245884.1
13
1504
XM_006710551.3
XP_006710614.1
14
1713
XM_011541179.2
XP_011539481.1
17
2083
XM_011541180.2
XP_011539482.1
17
2238
XM_011541181.1
XP_011539483.1
16
1912
XM_011541185.2
XP_011539487.1
14
1631
XM_011541186.2
XP_011539488.1
14
1728
XM_017000913.1
XP_016856402.1
13
1512
XM_017000914.1
XP_016856403.1
16
2132
XM_017000915.1
XP_016856404.1
16
1854
XM_017000916.1
XP_016856405.1
14
1766
XM_017000917.1
XP_016856406.1
15
1981
XM_017000918.1
XP_016856407.1
16
2178
XM_017000919.1
XP_016856408.1
15
1905
XM_017000920.1
XP_016856409.1
14
2123
XM_017000921.1
XP_016856410.1
15
1930
XM_017000922.1
XP_016856411.1
13
1589
GenBank mRNA
AK127543.1
-
3
385
AL832390.1
-
2
119
AX775875.1
-
13
1637
AX775877.1
-
12
1517
BC053600.1
-
12
1467
5744211
BX640987.1
-
14
1609
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0