Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206997.1 | NP_001193926.1 | 5, 6 | 577 | ||
NM_152770.2 | NP_689983.2 | 4, 5 | 526 | |||
XM_017007972.1 | XP_016863461.1 | 5, 6 | 678 | |||
XM_017007975.1 | XP_016863464.1 | 3, 4 | 666 | |||
XM_017007978.1 | XP_016863467.1 | 3, 4 | 543 | |||
GenBank mRNA | AY349359.1 | - | 5, 6 | 528 | ||
BC034296.1 | - | 4, 5 | 526 | 5165558 | ||
DB522959.1 | - | 1, 2 | 85 |