Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136885.1 | 4 | 318 | |||
GenBank mRNA | AK096042.1 | - | 4 | 318 | ||
BC131692.1 | - | 5 | 412 | 40119840 |