Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003936.3 | NP_001003936.1 | 2 | 158 | ||
NM_001286946.1 | NP_001273875.1 | 2 | 158 | |||
NM_001286947.1 | NP_001273876.1 | 2 | 158 | |||
XM_011518463.2 | XP_011516765.1 | 2 | 161 | |||
XM_017014579.1 | XP_016870068.1 | 2 | 161 | |||
XM_017014580.1 | XP_016870069.1 | 2 | 161 | |||
XM_017014581.1 | XP_016870070.1 | 2 | 161 | |||
XM_017014582.1 | XP_016870071.1 | 2 | 161 | |||
XM_017014583.1 | XP_016870072.1 | 2 | 161 | |||
XM_017014584.1 | XP_016870073.1 | 2 | 277 | |||
GenBank mRNA | AF305830.1 | - | 2 | 98 | ||
BC035743.1 | - | 2 | 119 | |||
BC130549.1 | - | 2 | 88 | 40146580 | ||
BC130551.1 | - | 2 | 88 | 40146582 | ||
BC144461.1 | - | 2 | 88 | 9052987 | ||
HY007335.1 | - | 2 | 159 |