Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039127.5 | NP_001034216.2 | 4 | 441 | ||
NM_001289930.1 | NP_001276859.1 | 4 | 1064 | |||
NM_001289931.1 | NP_001276860.1 | 4 | 366 | |||
NR_110529.1 | 2 | 218 | ||||
XM_017007979.1 | XP_016863468.1 | 5 | 2642 | |||
XM_017007980.1 | XP_016863469.1 | 5 | 2224 | |||
XM_017007981.1 | XP_016863470.1 | 5 | 471 | |||
GenBank mRNA | AK096662.1 | - | 3 | 1096 | ||
AL833758.1 | - | 2 | 174 | |||
BC047874.1 | - | 1 | 376 | |||
BC104026.1 | - | 4 | 1002 | 40025602 | ||
BC104027.2 | - | 4 | 379 | 40025603 | ||
BC104028.1 | - | 4 | 1002 | 40025604 | ||
BC104029.2 | - | 4 | 379 | 40025606 | ||
BC154406.1 | - | 4 | 379 | 40112082 | ||
CN265977.1 | - | 4 | 368 |