Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145411.1 | NP_001138883.1 | 5 | 1260 | ||
NM_152775.3 | NP_689988.1 | 6 | 1371 | |||
XM_005262891.3 | XP_005262948.1 | 6 | 1364 | |||
XM_006714172.2 | XP_006714235.1 | 6 | 1364 | |||
XM_011531827.2 | XP_011530129.1 | 6 | 1364 | |||
XM_011531828.2 | XP_011530130.1 | 6 | 1441 | |||
XM_011531829.2 | XP_011530131.1 | 5 | 1308 | |||
XM_017007987.1 | XP_016863476.1 | 5 | 1349 | |||
XM_017007988.1 | XP_016863477.1 | 5 | 1308 | |||
GenBank mRNA | AB080722.1 | - | 6 | 1337 | ||
AK097605.1 | - | 6 | 1364 | |||
AK302815.1 | - | 6 | 1310 | |||
AL833337.1 | - | 6 | 1316 | |||
BC028419.1 | - | 6 | 1336 | 4825416 | ||
BC038515.1 | - | 5 | 1200 | 5266693 | ||
BU567268.1 | - | 1 | 297 | |||
BX641119.1 | - | 6 | 1342 |