Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042683.2 | NP_001036148.2 | 5 | 1434 | ||
NM_173082.3 | NP_775105.1 | 5 | 1434 | |||
XM_006715439.3 | XP_006715502.1 | 5 | 1347 | |||
XM_006715441.3 | XP_006715504.1 | 5 | 1347 | |||
XM_006715443.3 | XP_006715506.1 | 5 | 1347 | |||
XM_011535719.2 | XP_011534021.1 | 5 | 1347 | |||
XM_011535722.2 | XP_011534024.1 | 5 | 1347 | |||
XM_017010691.1 | XP_016866180.1 | 5 | 1342 | |||
XM_017010692.1 | XP_016866181.1 | 5 | 1434 | |||
XM_017010693.1 | XP_016866182.1 | 5 | 1347 | |||
XM_017010694.1 | XP_016866183.1 | 5 | 943 | |||
XM_017010695.1 | XP_016866184.1 | 4 | 810 | |||
XM_017010696.1 | XP_016866185.1 | 5 | 896 | |||
GenBank mRNA | AB095943.1 | - | 4 | 969 | ||
AK075318.1 | - | 5 | 1433 | |||
AY161136.1 | - | 5 | 1386 | |||
AY163808.1 | - | 5 | 1433 | |||
BC117686.1 | - | 5 | 1100 | 40071177 | ||
BX648322.1 | - | 4 | 567 | |||
CR749290.1 | - | 6 | 1001 |