Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EXT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000401.3 | NP_000392.3 | 5 | 999 | ||
NM_001178083.1 | NP_001171554.1 | 5 | 1178 | |||
NM_207122.1 | NP_997005.1 | 5 | 1178 | |||
XM_011519950.1 | XP_011518252.1 | 6 | 997 | |||
XM_011519951.1 | XP_011518253.1 | 6 | 1107 | |||
GenBank mRNA | AK296713.1 | - | 5 | 692 | ||
AK312375.1 | - | 5 | 1055 | |||
BC010058.1 | - | 5 | 1052 | 3947503 | ||
BC068545.1 | - | 5 | 1018 | 5263866 | ||
BX648142.1 | - | 5 | 976 | |||
U62740.1 | - | 5 | 1178 | |||
U64511.1 | - | 4 | 870 | |||
U72263.1 | - | 5 | 1010 |