Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDCSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152997.3 | NP_694542.1 | 4 | 252 | ||
GenBank mRNA | AF086120.1 | - | 4 | 181 | ||
AF435080.1 | - | 4 | 188 | |||
AY190326.1 | - | 4 | 252 | |||
AY358958.1 | - | 4 | 225 | |||
BC062213.1 | - | 4 | 203 | 30326274 |