Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDCSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152997.3 | NP_694542.1 | 3, 4 | 206 | ||
GenBank mRNA | AF086120.1 | - | 3, 4 | 135 | ||
AF435080.1 | - | 3, 4 | 142 | |||
AY190326.1 | - | 3, 4 | 206 | |||
AY358958.1 | - | 3, 4 | 179 | |||
BC062213.1 | - | 3, 4 | 157 | 30326274 |