Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153615.1 | NP_705843.1 | 2 | 1426 | ||
GenBank mRNA | AY101396.1 | - | 2 | 1426 | ||
BC101108.3 | - | 1 | 1106 | 40011557 | ||
BC101109.2 | - | 1 | 1106 | 40011564 | ||
BC101110.3 | - | 2 | 981 |