Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025048.3 | NP_079324.2 | 5 | 528 | ||
NM_153840.3 | NP_722582.2 | 5 | 528 | |||
XM_011514469.1 | XP_011512771.1 | 3 | 243 | |||
XM_017010735.1 | XP_016866224.1 | 3 | 345 | |||
XM_017010736.1 | XP_016866225.1 | 3 | 261 | |||
GenBank mRNA | AB045123.1 | - | 5 | 482 | ||
AK026337.1 | - | 5 | 459 | |||
AK095348.1 | - | 3 | 345 | |||
AK225460.1 | - | 5 | 459 | |||
BC029358.1 | - | 4 | 353 | |||
BC104214.1 | - | 4 | 377 | |||
BC104215.2 | - | 3 | 265 | |||
BC113634.1 | - | 4 | 341 | |||
BC113636.1 | - | 4 | 341 | |||
CR627234.1 | - | 4 | 365 | |||
DB224175.1 | - | 5 | 530 |