Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199777.1 | NP_001186706.1 | 6 | 1064 | ||
NM_172240.2 | NP_758440.1 | 7 | 1083 | |||
NR_037659.1 | 6 | 911 | ||||
NR_037660.1 | 5 | 892 | ||||
GenBank mRNA | AB053266.1 | - | 6 | 573 | ||
AK027829.1 | - | 3 | 282 | |||
AK074772.1 | - | 7 | 821 | |||
AK123106.1 | - | 6 | 1061 | |||
AK295596.1 | - | 6 | 649 | |||
AL832918.1 | - | 5 | 548 | |||
BC026080.2 | - | 7 | 845 | 4827689 | ||
BU521431.1 | - | 5 | 561 | |||
EF011619.1 | - | 7 | 845 |