Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199777.1 | NP_001186706.1 | 10 | 1521 | ||
NM_172240.2 | NP_758440.1 | 11 | 1540 | |||
NR_037659.1 | 11 | 1391 | ||||
NR_037660.1 | 9 | 1349 | ||||
GenBank mRNA | AB053266.1 | - | 10 | 1030 | ||
AK027829.1 | - | 7 | 739 | |||
AK074772.1 | - | 11 | 1278 | |||
AK123106.1 | - | 10 | 1518 | |||
AK123625.1 | - | 7 | 738 | |||
AK295596.1 | - | 11 | 1129 | |||
AL832918.1 | - | 10 | 1027 | |||
BC026080.2 | - | 11 | 1302 | 4827689 | ||
BX648293.1 | - | 1 | 4142 | |||
EF011619.1 | - | 11 | 1302 |