Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK32C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173575.3 | NP_775846.2 | 1 | 384 | ||
XM_011539690.2 | XP_011537992.1 | 1 | 355 | |||
GenBank mRNA | AY098866.1 | - | 1 | 384 |