Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001342.2 | NP_001001342.1 | 2, 3 | 380 | ||
NM_001282436.1 | NP_001269365.1 | 4, 5 | 445 | |||
NM_001282437.1 | NP_001269366.1 | 3, 4 | 456 | |||
NM_001282438.1 | NP_001269367.1 | 3, 4 | 307 | |||
NM_001282439.1 | NP_001269368.1 | 3, 4 | 337 | |||
NM_173809.4 | NP_776170.2 | 3, 4 | 337 | |||
XM_017016102.1 | XP_016871591.1 | 3, 4 | 436 | |||
GenBank mRNA | AB593084.1 | - | 3, 4 | 299 | ||
AK054697.1 | - | 2, 3 | 373 | |||
AK298969.1 | - | 3, 4 | 305 | |||
AY278457.1 | - | 2, 3 | 335 | |||
AY531265.1 | - | 3, 4 | 284 | |||
BC020494.1 | - | 2, 3 | 208 | 3900723 | ||
BF666010.1 | - | 4, 5 | 414 | |||
BG545838.1 | - | 2, 3 | 375 | |||
BG679165.1 | - | 3, 4 | 309 | |||
BI753469.1 | - | 3, 4 | 297 | |||
CN340962.1 | - | 3, 4 | 424 | |||
DB456773.1 | - | 3, 4 | 305 | |||
HY107885.1 | - | 2, 3 | 382 | |||
HY165508.1 | - | 3, 4 | 308 |