Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145018.1 | NP_001138490.1 | 6 | 550 | ||
XM_006718818.3 | XP_006718881.1 | 3 | 430 | |||
XM_011542763.2 | XP_011541065.1 | 9 | 1037 | |||
XM_011542764.2 | XP_011541066.1 | 9 | 1058 | |||
XM_011542770.2 | XP_011541072.1 | 7 | 746 | |||
XM_011542771.2 | XP_011541073.1 | 5 | 716 | |||
XM_011542772.2 | XP_011541074.1 | 5 | 531 | |||
XM_011542774.2 | XP_011541076.1 | 8 | 881 | |||
XM_017017577.1 | XP_016873066.1 | 4 | 767 | |||
XM_017017578.1 | XP_016873067.1 | 7 | 2024 | |||
XM_017017579.1 | XP_016873068.1 | 4 | 315 | |||
XM_017017580.1 | XP_016873069.1 | 7 | 699 | |||
GenBank mRNA | BC043548.1 | - | 8 | 1622 | ||
BC101443.1 | - | 2 | 424 | 40022161 | ||
BC101444.1 | - | 2 | 424 | 40022159 | ||
BC101986.1 | - | 2 | 424 | 40022155 |