Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD163L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297650.1 | NP_001284579.1 | 12 | 3125 | ||
NM_174941.5 | NP_777601.2 | 12 | 3095 | |||
XM_011520616.1 | XP_011518918.1 | 12 | 3063 | |||
XM_011520617.2 | XP_011518919.1 | 11 | 3072 | |||
XM_011520618.2 | XP_011518920.1 | 10 | 2391 | |||
XM_011520619.1 | XP_011518921.1 | 11 | 2583 | |||
XM_011520620.1 | XP_011518922.1 | 10 | 2537 | |||
XM_011520621.2 | XP_011518923.1 | 5 | 1076 | |||
GenBank mRNA | AF264014.1 | - | 12 | 3062 | ||
AK058038.1 | - | 4 | 732 | |||
AK303372.1 | - | 12 | 3086 | |||
BC104962.1 | - | 12 | 3067 | 8143965 | ||
BC104964.1 | - | 12 | 3067 | 8143967 | ||
GQ397482.1 | - | 12 | 3037 |