Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD163L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297650.1 | NP_001284579.1 | 6 | 1268 | ||
NM_174941.5 | NP_777601.2 | 6 | 1238 | |||
XM_011520616.1 | XP_011518918.1 | 6 | 1206 | |||
XM_011520617.2 | XP_011518919.1 | 5 | 1215 | |||
XM_011520618.2 | XP_011518920.1 | 4 | 534 | |||
XM_011520619.1 | XP_011518921.1 | 5 | 726 | |||
XM_011520620.1 | XP_011518922.1 | 4 | 680 | |||
GenBank mRNA | AF264014.1 | - | 6 | 1205 | ||
AK303372.1 | - | 6 | 1229 | |||
BC104962.1 | - | 6 | 1210 | 8143965 | ||
BC104964.1 | - | 6 | 1210 | 8143967 | ||
GQ397482.1 | - | 6 | 1180 |