Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198827.3 | NP_942122.2 | 4 | 777 | ||
XM_005253566.2 | XP_005253623.1 | 4 | 777 | |||
XM_011538203.2 | XP_011536505.1 | 5 | 873 | |||
XM_011538204.2 | XP_011536506.1 | 5 | 873 | |||
XM_011538205.2 | XP_011536507.1 | 5 | 873 | |||
XM_011538206.2 | XP_011536508.1 | 5 | 873 | |||
XM_011538207.2 | XP_011536509.1 | 5 | 873 | |||
XM_011538208.2 | XP_011536510.1 | 5 | 873 | |||
XM_011538209.2 | XP_011536511.1 | 5 | 873 | |||
XM_011538210.2 | XP_011536512.1 | 5 | 873 | |||
GenBank mRNA | AY532280.1 | - | 4 | 391 | ||
BC112307.1 | - | 4 | 248 | 8327775 | ||
BC112309.1 | - | 4 | 248 | 8327777 | ||
BC143775.1 | - | 5 | 344 | 9052289 | ||
BX649116.1 | - | 3 | 5688 | |||
EU523118.1 | - | 4 | 292 |
Set Membership: |
![]() |