Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036486.1 | 3 | 398 | |||
GenBank mRNA | AK093430.1 | - | 2 | 301 | ||
AK125748.1 | - | 3 | 398 |
Gene Symbol: FGF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321946.1 | NP_001308875.1 | 3 | 529 | ||
NM_001321949.1 | NP_001308878.1 | 2 | 1116 |