Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 27, 28 | 3427 | ||
NM_173636.4 | NP_775907.4 | 27, 28 | 3412 | |||
XM_005258809.2 | XP_005258866.1 | 26, 27 | 3316 | |||
XM_011526837.1 | XP_011525139.1 | 27, 28 | 3412 | |||
XM_011526838.1 | XP_011525140.1 | 25, 26 | 3178 | |||
XM_011526839.1 | XP_011525141.1 | 25, 26 | 3076 | |||
XM_011526840.2 | XP_011525142.1 | 20, 21 | 2422 | |||
XM_011526841.2 | XP_011525143.1 | 18, 19 | 2043 | |||
XM_011526842.1 | XP_011525144.1 | 17, 18 | 1875 | |||
XM_011526843.1 | XP_011525145.1 | 10, 11 | 1202 | |||
XM_011526844.2 | XP_011525146.1 | 10, 11 | 1199 | |||
XM_017026665.1 | XP_016882154.1 | 27, 28 | 3427 | |||
GenBank mRNA | AL117438.1 | - | 1, 2 | 117 | ||
AL133651.1 | - | 6, 7 | 729 | |||
BC017261.2 | - | 9, 10 | 1121 | |||
BC058939.1 | - | 27, 28 | 3371 | |||
BX647726.1 | - | 25, 26 | 4658 | |||
KU178897.1 | - | 6, 7 | 657 | |||
KU178898.1 | - | 6, 7 | 672 | |||
KU178899.1 | - | 4, 5 | 423 |