Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532783.1 | XP_011531085.1 | 11 | 952 | ||
XM_011532796.2 | XP_011531098.1 | 7 | 373 | |||
XM_017003834.1 | XP_016859323.1 | 14 | 4833 | |||
XM_017003835.1 | XP_016859324.1 | 14 | 4260 | |||
XM_017003836.1 | XP_016859325.1 | 13 | 4584 | |||
XM_017003837.1 | XP_016859326.1 | 14 | 4702 | |||
XM_017003838.1 | XP_016859327.1 | 13 | 4321 | |||
XM_017003839.1 | XP_016859328.1 | 13 | 1876 | |||
XM_017003840.1 | XP_016859329.1 | 13 | 4932 | |||
XM_017003841.1 | XP_016859330.1 | 13 | 4954 | |||
XM_017003842.1 | XP_016859331.1 | 15 | 4681 | |||
XM_017003843.1 | XP_016859332.1 | 14 | 4566 | |||
XM_017003844.1 | XP_016859333.1 | 15 | 3892 | |||
XM_017003845.1 | XP_016859334.1 | 15 | 4334 | |||
XM_017003846.1 | XP_016859335.1 | 15 | 4627 | |||
XM_017003849.1 | XP_016859338.1 | 14 | 4612 | |||
XM_017003850.1 | XP_016859339.1 | 14 | 4512 | |||
XM_017003851.1 | XP_016859340.1 | 12 | 4873 | |||
XM_017003852.1 | XP_016859341.1 | 11 | 537 | |||
XM_017003859.1 | XP_016859348.1 | 11 | 673 | |||
XM_017003860.1 | XP_016859349.1 | 8 | 415 | |||
GenBank mRNA | AF450483.1 | - | 2 | 402 | ||
BC110073.1 | - | 10 | 614 | |||
DB507279.3 | - | 4 | 207 |