Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 27 | 3254 | ||
XM_006712438.3 | XP_006712501.1 | 26 | 3191 | |||
XM_006712439.3 | XP_006712502.1 | 11 | 1838 | |||
XM_011510985.2 | XP_011509287.1 | 25 | 2983 | |||
XM_011510987.2 | XP_011509289.1 | 19 | 2174 | |||
XM_011510988.2 | XP_011509290.1 | 23 | 2700 | |||
XM_017003861.1 | XP_016859350.1 | 25 | 3098 | |||
XM_017003862.1 | XP_016859351.1 | 25 | 3077 | |||
XM_017003863.1 | XP_016859352.1 | 25 | 2996 | |||
XM_017003865.1 | XP_016859354.1 | 23 | 2715 | |||
XM_017003866.1 | XP_016859355.1 | 20 | 2390 | |||
GenBank mRNA | AK001064.1 | - | 4 | 1383 | ||
AK125768.1 | - | 11 | 2222 | |||
AK302685.1 | - | 22 | 2622 | |||
AL834537.1 | - | 10 | 2138 |