Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207351.4 | NP_997234.3 | 4 | 1718 | ||
NR_134900.1 | 4 | 1718 | ||||
XM_011533628.2 | XP_011531930.1 | 4 | 1796 | |||
XM_011533629.2 | XP_011531931.1 | 4 | 1776 | |||
XM_017006256.1 | XP_016861745.1 | 3 | 3583 | |||
GenBank mRNA | AK090993.1 | - | 4 | 1697 | ||
BC040508.1 | - | 4 | 1695 | 5300209 | ||
BC089447.1 | - | 4 | 1677 | 30563721 |