Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297732.1 | NP_001284661.1 | 5 | 872 | ||
NM_001297733.1 | NP_001284662.1 | 5 | 873 | |||
NM_001300729.1 | NP_001287658.1 | 4 | 768 | |||
NM_173827.3 | NP_776188.1 | 5 | 869 | |||
XM_005265680.4 | XP_005265737.1 | 5 | 872 | |||
XM_011531878.2 | XP_011530180.1 | 5 | 843 | |||
XM_017008045.1 | XP_016863534.1 | 5 | 869 | |||
GenBank mRNA | AK096310.2 | - | 5 | 815 | ||
AM055750.1 | - | 5 | 519 | |||
AM055751.1 | - | 5 | 523 | |||
AM055752.1 | - | 4 | 418 | |||
AY957564.1 | - | 5 | 865 | |||
AY957565.1 | - | 5 | 865 | |||
BC101684.1 | - | 5 | 802 | 8069190 | ||
BC143642.1 | - | 5 | 805 | 9052156 |