Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174952.2 | NP_777612.1 | 8, 9 | 1127 | ||
XM_011531886.2 | XP_011530188.1 | 10, 11 | 1627 | |||
XM_017008049.1 | XP_016863538.1 | 8, 9 | 1402 | |||
XM_017008050.1 | XP_016863539.1 | 8, 9 | 1360 | |||
XM_017008051.1 | XP_016863540.1 | 5, 6 | 605 | |||
GenBank mRNA | BC036870.1 | - | 8, 9 | 1127 | 5226859 | |
BX648305.1 | - | 2, 3 | 179 |